Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 7 - 8 | 972 | 90 | |
| GenBank mRNA | AK159104.1 | - | 7 - 8 | 973 | 90 | |
| BC039963.1 | - | 5 - 6 | 758 | 4240646 | 90 | |
| BC061234.1 | - | 6 - 7 | 811 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhps
Genomic Map