Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 8 - 9 | 1097 | 84 | |
| GenBank mRNA | BB763715.1 | - | 3 - 4 | 290 | 84 | |
| BC039963.1 | - | 6 - 7 | 883 | 4240646 | 84 | |
| BC061234.1 | - | 7 - 8 | 936 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023553 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040280 | NM_001039514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040280 | AK159104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040280 | AK177102.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040280 | AK185697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031112 | BC039963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031112 | BC061234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031112 | NM_001039514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031112 | AK159104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031112 | AK212076.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429626 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136720 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031112 | BC039963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031112 | BC061234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031112 | NM_001039514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031112 | AK159104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031112 | AK212076.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040280 | NM_001039514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040280 | AK159104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040280 | AK177102.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040280 | AK185697.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023538 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136720 | - | - |
| Whole Mouse Genome G4122A | A_51_P279851 | BC039963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279851 | BC061234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279851 | NM_001039514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279851 | AK159104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279851 | AK212076.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136720 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07291452 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06726658 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07291448 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07182162 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07291441 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06726681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07291446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07291443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06726673 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06679614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06726681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07182162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06679626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06679621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06726667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07306964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06726673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06679615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573572 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434003_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434004_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573566 | - | - |
| Mouse Genome 430 2.0 Array | 1434004_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434003_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434004_at | - | - |
| Mouse Genome 430 2.0 Array | 1434003_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330817_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330817_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800007542.mm.1 | NM_001039514.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007542.mm.1 | NM_001039514.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007542.mm.1 | TC0800000981.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434004_at | - | - |
| Mouse Expression Set 430 | 1434003_PM_a_at | - | - |
| Mouse Expression Set 430 | 1434003_a_at | - | - |
| Mouse Expression Set 430 | 1434004_PM_at | - | - |
| Murine Genome U74v2 Set | 160405_at | - | - |
| Murine Genome U74v2 Set | 160405_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434003_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434004_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434003_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434004_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434004_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434003_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434004_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434003_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 842381 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637241 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2637241 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2637241 | - | - |

Dhps
Genomic Map