Assay Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019652.1 | NP_062626.1 | 2 - 3 | 322 | 107 | |
| GenBank mRNA | AF039405.2 | - | 2 - 3 | 322 | 107 | |
| AK007827.1 | - | 2 - 3 | 325 | 107 | ||
| AK171690.1 | - | 2 - 3 | 326 | 107 | ||
| AK192795.1 | - | 1 - 2 | 117 | 107 | ||
| AK216123.1 | - | 1 - 2 | 117 | 107 | ||
| BC016453.1 | - | 2 - 3 | 310 | 4920063 | 107 | |
| BC083335.1 | - | 2 - 3 | 320 | 5717763 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007827.1 transcripts.
Set Membership: |
|

Asna1
Genomic Map