Assay Details
| Gene Symbol: Gpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173866.3 | NP_776291.1 | 2 - 3 | 337 | 64 | |
| GenBank mRNA | AK031958.1 | - | 2 - 3 | 349 | 64 | |
| AK033424.1 | - | 2 - 3 | 338 | 64 | ||
| AK075894.1 | - | 2 - 3 | 356 | 64 | ||
| AK076250.1 | - | 2 - 3 | 355 | 64 | ||
| AK082030.1 | - | 2 - 3 | 380 | 64 | ||
| BC034219.1 | - | 2 - 3 | 369 | 5065322 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpt2
Genomic Map