Assay Details
| Gene Symbol: Orc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019716.2 | NP_062690.2 | 1 - 2 | 355 | 115 | |
| NR_028268.1 | 1 - 2 | 355 | 115 | |||
| GenBank mRNA | AF139659.1 | - | 1 - 2 | 196 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF139659.1, NM_019716.2 transcripts.
Set Membership: |
|

Orc6
Genomic Map