Assay Details
| Gene Symbol: Orc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163791.1 | NP_001157263.1 | 5 - 6 | 826 | 141 | |
| NM_019716.2 | NP_062690.2 | 6 - 7 | 956 | 141 | ||
| NR_028268.1 | 5 - 6 | 843 | 141 | |||
| GenBank mRNA | AF139659.1 | - | 6 - 7 | 797 | 141 | |
| AK078405.1 | - | 5 - 6 | 560 | 141 | ||
| AK163256.1 | - | 5 - 6 | 770 | 141 | ||
| BC048689.1 | - | 5 - 6 | 552 | 141 | ||
| BC080739.1 | - | 6 - 7 | 675 | 30459034 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024227 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171253 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125098 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26024229 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125098 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06580474 | - | - |
| Whole Mouse Genome G4122A | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | AK078405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | AK163256.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BC048689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BC080739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296416 | BP771997.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123240 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06674292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06674276 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06674286 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06674283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26024228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26024272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07274014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06674276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06951984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07005339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07274012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06748181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06674283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06951982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26024234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573621 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417037_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573615 | - | - |
| Mouse Genome 430 2.0 Array | 1417037_at | - | - |
| Mouse Genome 430 2.0 Array | 1417037_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56452_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56452_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NM_001163791.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NM_001163791.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NM_019716.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NM_019716.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NR_028268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | NR_028268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007599.mm.1 | TC0800000987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NM_001163791.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NM_001163791.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NM_019716.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NM_019716.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NR_028268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | NR_028268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007598.mm.1 | TC0800000987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NM_001163791.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NM_001163791.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NM_019716.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NM_019716.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NR_028268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | NR_028268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007600.mm.1 | TC0800000987.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417037_PM_at | - | - |
| Mouse Expression Set 430 | 1417037_at | - | - |
| Murine Genome U74v2 Set | 95712_at | - | - |
| Murine Genome U74v2 Set | 95712_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417037_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417037_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417037_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417037_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 923925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683190 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2683190 | - | - |
| mouse-6 v1.1 (Illumina) | scl33558.9.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33558.9.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2683190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683190 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33558.9.1_16-S | - | - |

Orc6
Genomic Map