Assay Details
| Gene Symbol: Hook2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167991.1 | NP_001161463.1 | 14 - 15 | 1486 | 77 | |
| NM_133255.2 | NP_573518.2 | 14 - 15 | 1486 | 77 | ||
| GenBank mRNA | AK030599.1 | - | 14 - 15 | 1479 | 77 | |
| AK042048.1 | - | 14 - 15 | 1474 | 77 | ||
| AK136069.1 | - | 15 - 16 | 1408 | 77 | ||
| AK145768.1 | - | 14 - 15 | 1454 | 77 | ||
| AK161612.1 | - | 14 - 15 | 1439 | 77 | ||
| AK166954.1 | - | 14 - 15 | 1466 | 77 | ||
| AK169073.1 | - | 14 - 15 | 1454 | 77 | ||
| BC002226.1 | - | 6 - 7 | 695 | 77 | ||
| BC019486.1 | - | 14 - 15 | 1462 | 4211990 | 77 | |
| BC026609.1 | - | 6 - 7 | 646 | 77 | ||
| BC029967.1 | - | 14 - 15 | 1427 | 77 | ||
| BC055881.1 | - | 14 - 15 | 1432 | 3590336 | 77 | |
| BC080744.1 | - | 14 - 15 | 1476 | 30471369 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook2
Genomic Map