Assay Details
| Gene Symbol: Hook2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167991.1 | NP_001161463.1 | 17 - 18 | 1743 | 57 | |
| NM_133255.2 | NP_573518.2 | 17 - 18 | 1743 | 57 | ||
| GenBank mRNA | AK042048.1 | - | 17 - 18 | 1731 | 57 | |
| AK136069.1 | - | 18 - 19 | 1665 | 57 | ||
| AK145768.1 | - | 17 - 18 | 1711 | 57 | ||
| AK166954.1 | - | 17 - 18 | 1723 | 57 | ||
| AK169073.1 | - | 17 - 18 | 1711 | 57 | ||
| BC002226.1 | - | 9 - 10 | 952 | 57 | ||
| BC019486.1 | - | 17 - 18 | 1719 | 4211990 | 57 | |
| BC026609.1 | - | 9 - 10 | 903 | 57 | ||
| BC029967.1 | - | 17 - 18 | 1684 | 57 | ||
| BC055881.1 | - | 17 - 18 | 1689 | 3590336 | 57 | |
| BC080744.1 | - | 17 - 18 | 1733 | 30471369 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook2
Genomic Map