Assay Details
| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146059.1 | NP_001139531.1 | 11 - 12 | 1304 | 57 | |
| NM_001146060.1 | NP_001139532.1 | 11 - 12 | 1384 | 57 | ||
| NM_146228.4 | NP_666340.3 | 11 - 12 | 1301 | 57 | ||
| GenBank mRNA | AB107016.1 | - | 11 - 12 | 1301 | 57 | |
| AF481142.1 | - | 10 - 11 | 1197 | 57 | ||
| AK030815.1 | - | 4 - 5 | 438 | 57 | ||
| AK053076.1 | - | 11 - 12 | 1274 | 57 | ||
| AK131149.1 | - | 11 - 12 | 1303 | 57 | ||
| AK155451.1 | - | 11 - 12 | 1370 | 57 | ||
| BC117928.1 | - | 10 - 11 | 1254 | 40097635 | 57 | |
| BC117929.1 | - | 10 - 11 | 1251 | 40097636 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030815.1 transcripts.
Set Membership: |
|

Als2cl
Genomic Map