Assay Details
| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146059.1 | NP_001139531.1 | 20 - 21 | 2377 | 99 | |
| NM_001146060.1 | NP_001139532.1 | 20 - 21 | 2457 | 99 | ||
| NM_146228.4 | NP_666340.3 | 20 - 21 | 2374 | 99 | ||
| GenBank mRNA | AB107016.1 | - | 20 - 21 | 2374 | 99 | |
| AF481142.1 | - | 19 - 20 | 2270 | 99 | ||
| AK030815.1 | - | 12 - 13 | 1414 | 99 | ||
| AK053076.1 | - | 20 - 21 | 2347 | 99 | ||
| AK131149.1 | - | 19 - 20 | 2279 | 99 | ||
| AK155451.1 | - | 20 - 21 | 2443 | 99 | ||
| BC022645.1 | - | 1 - 2 | 34 | 99 | ||
| BC117928.1 | - | 19 - 20 | 2327 | 40097635 | 99 | |
| BC117929.1 | - | 19 - 20 | 2324 | 40097636 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Als2cl
Genomic Map