| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012354.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012352.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012344.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012344.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012344.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012344.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012344.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012349.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012349.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012349.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012360.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012356.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012357.mm.1 |
TC0900001475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012358.mm.1 |
TC0900001475.mm.1 |
MTA TC |