Assay Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009204.2 | NP_033230.2 | 5 - 6 | 771 | 65 | |
| GenBank mRNA | AB008453.1 | - | 5 - 6 | 569 | 65 | |
| AK137607.1 | - | 5 - 6 | 756 | 65 | ||
| AK138832.1 | - | 5 - 6 | 745 | 65 | ||
| AK160882.1 | - | 4 - 5 | 872 | 65 | ||
| AK164310.1 | - | 5 - 6 | 746 | 65 | ||
| BC014282.1 | - | 5 - 6 | 741 | 4207674 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc2a4
Genomic Map