| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100028266.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028266.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028266.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028264.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028264.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028264.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028271.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028271.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028271.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028265.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028265.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028265.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028268.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028268.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028268.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028274.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028274.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028274.mm.1 |
TC1100003075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100028272.mm.1 |
NM_027419.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028272.mm.1 |
NM_027419.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100028272.mm.1 |
TC1100003075.mm.1 |
MTA TC |