Assay Details
| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164659.1 | NP_001158131.1 | 16 - 17 | 5476 | 116 | |
| GenBank mRNA | AK129119.1 | - | 4 - 5 | 1542 | 116 | |
| BC086653.1 | - | 4 - 5 | 502 | 30536238 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trank1
Genomic Map