Assay Details
| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164659.1 | NP_001158131.1 | 17 - 18 | 5571 | 89 | |
| GenBank mRNA | AK129119.1 | - | 5 - 6 | 1637 | 89 | |
| BC086653.1 | - | 5 - 6 | 597 | 30536238 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trank1
Genomic Map