Assay Details
| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164659.1 | NP_001158131.1 | 8 - 9 | 1431 | 96 | |
| GenBank mRNA | AK079552.1 | - | 8 - 9 | 1432 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trank1
Genomic Map