Assay Details
| Gene Symbol: Hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024219.1 | NP_077181.1 | 1 - 2 | 86 | 91 | |
| GenBank mRNA | AK010939.1 | - | 640 | 91 | ||
| AK018708.1 | - | 1 - 2 | 118 | 91 | ||
| AK050550.1 | - | 1 - 2 | 136 | 91 | ||
| AK078498.1 | - | 1 - 2 | 118 | 91 | ||
| AK145940.1 | - | 1 - 2 | 96 | 91 | ||
| BC002153.1 | - | 1 - 2 | 86 | 3485301 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsbp1
Genomic Map