Assay Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172561.3 | NP_766149.2 | 2 - 3 | 291 | 72 | |
| GenBank mRNA | AK042294.1 | - | 2 - 3 | 170 | 72 | |
| AK077543.1 | - | 2 - 3 | 169 | 72 | ||
| AK166184.1 | - | 2 - 3 | 170 | 72 | ||
| BC046751.1 | - | 2 - 3 | 152 | 72 | ||
| BC055821.1 | - | 2 - 3 | 158 | 4502461 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spag7
Genomic Map