Assay Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167669.1 | NP_001161141.1 | 6 - 7 | 678 | 61 | |
| NM_172561.3 | NP_766149.2 | 6 - 7 | 708 | 61 | ||
| GenBank mRNA | AK042294.1 | - | 6 - 7 | 587 | 61 | |
| AK077543.1 | - | 6 - 7 | 586 | 61 | ||
| AK166184.1 | - | 6 - 7 | 587 | 61 | ||
| BC046751.1 | - | 6 - 7 | 569 | 61 | ||
| BC055821.1 | - | 6 - 7 | 575 | 4502461 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902004 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102365 | AK166184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102365 | AK042294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102365 | AK077543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102365 | BC046751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102365 | BC055821.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53747 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102365 | AK166184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102365 | AK042294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102365 | AK077543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102365 | BC046751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102365 | BC055821.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07389223 | - | - |
| Whole Mouse Genome G4122A | A_51_P335583 | AK166184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335583 | AK042294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335583 | AK077543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335583 | BC046751.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335583 | BC055821.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127475 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07389223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09653250 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09653242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09661187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09079703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422117 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387943 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433940_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387936 | - | - |
| Mouse Genome 430 2.0 Array | 1433940_at | - | - |
| Mouse Genome 430 2.0 Array | 1452229_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452229_at | - | - |
| Mouse Genome 430 2.0 Array | 1433940_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216873_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216873_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100028705.mm.1 | NM_001167669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028705.mm.1 | NM_001167669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028705.mm.1 | NM_172561.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028705.mm.1 | NM_172561.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028705.mm.1 | TC1100003109.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028707.mm.1 | NM_001167669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028707.mm.1 | NM_001167669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028707.mm.1 | NM_172561.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028707.mm.1 | NM_172561.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028707.mm.1 | TC1100003109.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028704.mm.1 | NM_001167669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028704.mm.1 | NM_001167669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028704.mm.1 | NM_172561.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028704.mm.1 | NM_172561.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028704.mm.1 | TC1100003109.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433940_at | - | - |
| Mouse Expression Set 430 | 1433940_PM_at | - | - |
| Murine Genome U74v2 Set | 93832_at | - | - |
| Murine Genome U74v2 Set | 93832_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433940_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433940_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4560524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560524 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775089 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775089 | - | - |
| mouse-6 v1.1 (Illumina) | scl39991.5_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560524 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39991.5_43-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2830771 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775088 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2830771 | - | - |

Spag7
Genomic Map