Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 11 - 12 | 1971 | 76 | |
| NM_001291142.1 | NP_001278071.1 | 11 - 12 | 1980 | 76 | ||
| NM_001291143.1 | NP_001278072.1 | 13 - 14 | 2171 | 76 | ||
| NM_019400.3 | NP_062273.2 | 12 - 13 | 2100 | 76 | ||
| GenBank mRNA | AB001750.1 | - | 13 - 14 | 2007 | 76 | |
| AF248489.1 | - | 11 - 12 | 1766 | 76 | ||
| AF248490.1 | - | 11 - 12 | 1757 | 76 | ||
| AK134587.1 | - | 12 - 13 | 2101 | 76 | ||
| BC003921.1 | - | 3 - 4 | 721 | 76 | ||
| BC060166.1 | - | 12 - 13 | 2118 | 76 | ||
| BC080274.1 | - | 12 - 13 | 2072 | 76 | ||
| BC129853.1 | - | 12 - 13 | 1919 | 40109688 | 76 | |
| BC129854.1 | - | 12 - 13 | 1919 | 40109689 | 76 | |
| D86066.1 | - | 12 - 13 | 1936 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map