Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 14 - 15 | 2354 | 124 | |
| NM_001291142.1 | NP_001278071.1 | 14 - 15 | 2363 | 124 | ||
| NM_001291143.1 | NP_001278072.1 | 16 - 17 | 2554 | 124 | ||
| NM_019400.3 | NP_062273.2 | 15 - 16 | 2483 | 124 | ||
| GenBank mRNA | AB001750.1 | - | 16 - 17 | 2390 | 124 | |
| AF248489.1 | - | 14 - 15 | 2149 | 124 | ||
| AF248490.1 | - | 14 - 15 | 2140 | 124 | ||
| AK134587.1 | - | 15 - 16 | 2484 | 124 | ||
| BC003921.1 | - | 6 - 7 | 1104 | 124 | ||
| BC080274.1 | - | 15 - 16 | 2455 | 124 | ||
| BC129853.1 | - | 15 - 16 | 2308 | 40109688 | 130 | |
| BC129854.1 | - | 15 - 16 | 2302 | 40109689 | 124 | |
| D86066.1 | - | 15 - 16 | 2319 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map