Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 15 - 16 | 2450 | 122 | |
| NM_001291142.1 | NP_001278071.1 | 15 - 16 | 2459 | 122 | ||
| NM_001291143.1 | NP_001278072.1 | 17 - 18 | 2650 | 122 | ||
| NM_019400.3 | NP_062273.2 | 16 - 17 | 2579 | 122 | ||
| GenBank mRNA | AK134587.1 | - | 16 - 17 | 2580 | 122 | |
| BC003921.1 | - | 7 - 8 | 1200 | 122 | ||
| BC060166.1 | - | 15 - 16 | 3129 | 122 | ||
| BC080274.1 | - | 16 - 17 | 2551 | 122 | ||
| BC129853.1 | - | 16 - 17 | 2404 | 40109688 | 122 | |
| BC129854.1 | - | 16 - 17 | 2398 | 40109689 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map