Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 2 - 3 | 454 | 137 | |
| NM_001291142.1 | NP_001278071.1 | 3 - 4 | 583 | 137 | ||
| NM_001291143.1 | NP_001278072.1 | 4 - 5 | 654 | 137 | ||
| NM_019400.3 | NP_062273.2 | 3 - 4 | 583 | 137 | ||
| GenBank mRNA | AB001750.1 | - | 4 - 5 | 490 | 137 | |
| AF248489.1 | - | 3 - 4 | 369 | 137 | ||
| AF248490.1 | - | 2 - 3 | 240 | 137 | ||
| AK019413.1 | - | 3 - 4 | 557 | 137 | ||
| AK134587.1 | - | 3 - 4 | 582 | 137 | ||
| AK137236.1 | - | 3 - 4 | 548 | 137 | ||
| AK151901.1 | - | 3 - 4 | 552 | 137 | ||
| BC009814.1 | - | 3 - 4 | 579 | 137 | ||
| BC060166.1 | - | 3 - 4 | 562 | 137 | ||
| BC080274.1 | - | 3 - 4 | 556 | 137 | ||
| BC129853.1 | - | 3 - 4 | 402 | 40109688 | 137 | |
| BC129854.1 | - | 3 - 4 | 402 | 40109689 | 137 | |
| BY718394.1 | - | 3 - 4 | 137 | |||
| D86066.1 | - | 3 - 4 | 419 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map