Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 9 - 10 | 1745 | 113 | |
| NM_001291142.1 | NP_001278071.1 | 9 - 10 | 1754 | 113 | ||
| NM_001291143.1 | NP_001278072.1 | 11 - 12 | 1945 | 113 | ||
| NM_019400.3 | NP_062273.2 | 10 - 11 | 1874 | 113 | ||
| GenBank mRNA | AB001750.1 | - | 11 - 12 | 1781 | 113 | |
| AF248489.1 | - | 9 - 10 | 1540 | 113 | ||
| AF248490.1 | - | 9 - 10 | 1531 | 113 | ||
| AK134587.1 | - | 10 - 11 | 1875 | 113 | ||
| BC060166.1 | - | 10 - 11 | 1892 | 113 | ||
| BC080274.1 | - | 10 - 11 | 1846 | 113 | ||
| BC129853.1 | - | 10 - 11 | 1693 | 40109688 | 113 | |
| BC129854.1 | - | 10 - 11 | 1693 | 40109689 | 113 | |
| D86066.1 | - | 10 - 11 | 1710 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map