Assay Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153103.2 | NP_694743.2 | 14 - 15 | 1706 | 84 | |
| GenBank mRNA | AB074017.1 | - | 12 - 13 | 1336 | 84 | |
| AK046574.1 | - | 9 - 10 | 947 | 84 | ||
| AK173007.1 | - | 13 - 14 | 1413 | 84 | ||
| BC029286.1 | - | 14 - 15 | 1693 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif1c
Genomic Map