Assay Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153103.2 | NP_694743.2 | 6 - 7 | 797 | 67 | |
| GenBank mRNA | AB001456.1 | - | 2 - 3 | 148 | 67 | |
| AB074017.1 | - | 4 - 5 | 427 | 67 | ||
| AK029250.1 | - | 6 - 7 | 724 | 67 | ||
| AK046574.1 | - | 1 - 2 | 38 | 67 | ||
| AK173007.1 | - | 5 - 6 | 504 | 67 | ||
| BC029286.1 | - | 6 - 7 | 784 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif1c
Genomic Map