Assay Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083331.2 | NP_001076800.1 | 12 - 13 | 1808 | 103 | |
| NM_001276406.1 | NP_001263335.1 | 12 - 13 | 1808 | 103 | ||
| NM_172394.3 | NP_765982.2 | 12 - 13 | 1793 | 103 | ||
| NR_075087.1 | 11 - 12 | 2252 | 103 | |||
| GenBank mRNA | AJ532593.1 | - | 12 - 13 | 1802 | 103 | |
| AK002643.1 | - | 12 - 13 | 1794 | 103 | ||
| AK028563.1 | - | 12 - 13 | 1798 | 103 | ||
| AK085776.1 | - | 12 - 13 | 1792 | 103 | ||
| AK139738.1 | - | 11 - 12 | 2160 | 103 | ||
| AK160619.1 | - | 12 - 13 | 1794 | 103 | ||
| AK166214.1 | - | 12 - 13 | 1798 | 103 | ||
| BC032929.1 | - | 12 - 13 | 1774 | 1379673 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup88
Genomic Map