Assay Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291146.1 | NP_001278075.1 | 2 - 3 | 514 | 174 | |
| NM_001291147.1 | NP_001278076.1 | 2 - 3 | 258 | 174 | ||
| NM_033562.4 | NP_291040.1 | 2 - 3 | 223 | 174 | ||
| GenBank mRNA | AF208064.1 | - | 2 - 3 | 162 | 174 | |
| AK048627.1 | - | 2 - 3 | 170 | 174 | ||
| AK077659.1 | - | 2 - 3 | 174 | 174 | ||
| AK082564.1 | - | 2 - 3 | 183 | 174 | ||
| AK083632.1 | - | 2 - 3 | 162 | 174 | ||
| AK090105.1 | - | 2 - 3 | 233 | 174 | ||
| AK151934.1 | - | 2 - 3 | 514 | 174 | ||
| BC005682.1 | - | 2 - 3 | 162 | 3154448 | 174 | |
| BE627680.1 | - | 2 - 3 | 191 | 174 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151934.1, NM_001291146.1, NM_001291147.1 transcripts.
Set Membership: |
|

Derl2
Genomic Map