Assay Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291146.1 | NP_001278075.1 | 4 - 5 | 681 | 123 | |
| NM_001291147.1 | NP_001278076.1 | 4 - 5 | 425 | 123 | ||
| NM_001291148.1 | NP_001278077.1 | 4 - 5 | 387 | 123 | ||
| NM_033562.4 | NP_291040.1 | 4 - 5 | 390 | 123 | ||
| GenBank mRNA | AF208064.1 | - | 4 - 5 | 329 | 123 | |
| AK048627.1 | - | 4 - 5 | 337 | 123 | ||
| AK077659.1 | - | 4 - 5 | 341 | 123 | ||
| AK082564.1 | - | 4 - 5 | 350 | 123 | ||
| AK083632.1 | - | 4 - 5 | 329 | 123 | ||
| AK090105.1 | - | 4 - 5 | 400 | 122 | ||
| AK151934.1 | - | 4 - 5 | 681 | 123 | ||
| BC005682.1 | - | 4 - 5 | 329 | 3154448 | 123 | |
| BE627680.1 | - | 4 - 5 | 358 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Derl2
Genomic Map