Assay Details
Gene Symbol: Derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291146.1 | NP_001278075.1 | 5 - 6 | 879 | 124 | |
NM_001291147.1 | NP_001278076.1 | 5 - 6 | 623 | 124 | ||
NM_001291148.1 | NP_001278077.1 | 5 - 6 | 585 | 124 | ||
NM_033562.4 | NP_291040.1 | 5 - 6 | 588 | 124 | ||
GenBank mRNA | AF208064.1 | - | 5 - 6 | 527 | 124 | |
AK048627.1 | - | 5 - 6 | 535 | 124 | ||
AK077659.1 | - | 5 - 6 | 539 | 124 | ||
AK083632.1 | - | 5 - 6 | 527 | 124 | ||
AK083725.1 | - | 2 - 3 | 3291 | 124 | ||
AK151934.1 | - | 5 - 6 | 879 | 124 | ||
BC005682.1 | - | 5 - 6 | 527 | 3154448 | 124 | |
BC037503.1 | - | 1 - 2 | 170 | 124 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
