Assay Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291147.1 | NP_001278076.1 | 1 - 2 | 196 | 109 | |
| GenBank mRNA | AK090105.1 | - | 1 - 2 | 171 | 109 | |
| BE627680.1 | - | 1 - 2 | 129 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291147.1 transcripts.
Set Membership: |
|

Derl2
Genomic Map