Assay Details
Gene Symbol: Tmie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146260.2 | NP_666372.1 | 2 - 3 | 1028 | 115 | |
GenBank mRNA | AF481143.1 | - | 2 - 3 | 1028 | 115 | |
AK030341.1 | - | 2 - 3 | 1058 | 115 | ||
BC138840.1 | - | 2 - 3 | 592 | 8861862 | 115 | |
BC141124.1 | - | 2 - 3 | 592 | 9055736 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF481143.1, BC138840.1, BC141124.1, NM_146260.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
