Assay Details
| Gene Symbol: Xaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037713.4 | NP_001032802.2 | 4 - 5 | 497 | 94 | |
| GenBank mRNA | AK046736.1 | - | 3 - 4 | 268 | 94 | |
| CK130151.1 | - | 3 - 4 | 432 | 94 | ||
More Information
Set Membership: |
|

Xaf1
Genomic Map