Assay Details
| Gene Symbol: Fam110a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289150.1 | NP_001276079.1 | 1 - 2 | 79 | 64 | |
| GenBank mRNA | BC018162.1 | - | 1 - 2 | 64 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289150.1 transcripts.
Set Membership: |
|

Fam110a
Genomic Map