Assay Details
| Gene Symbol: Fam110a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080248.1 | - | 368 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
R-spondin family, member 4 |
Rspo4 |
A730099F22, A930029K19Rik |
Anti-sense | Chr. 2 - 151842579 - 151874668 on NCBI Build GRCm38 | |
| Downstream | angiopoietin 4 | Angpt4 |
Agpt4, Ang3 |
44.4kb | Anti-sense | Chr. 2 - 151911332 - 151944654 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Fam110a
Genomic Map