Assay Details
| Gene Symbol: Rnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK075970.1 | - | 407 | 158 | ||
| AK170529.1 | - | 645 | 158 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075970.1 transcripts.
Set Membership: |
|

Rnd2
Genomic Map