Assay Details
| Gene Symbol: Rnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009708.1 | NP_033838.1 | 2 - 3 | 188 | 62 | |
| GenBank mRNA | AF016482.1 | - | 2 - 3 | 171 | 62 | |
| AK075970.1 | - | 496 | 62 | |||
| BC107022.2 | - | 2 - 3 | 187 | 62 | ||
| BC127034.1 | - | 2 - 3 | 182 | 62 | ||
| BC128475.1 | - | 2 - 3 | 187 | 62 | ||
| BC139330.1 | - | 2 - 3 | 261 | 8862352 | 62 | |
| BC139331.1 | - | 2 - 3 | 261 | 8862353 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075970.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnd2
Genomic Map