Assay Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016695.3 | NP_057904.1 | 10 - 11 | 1275 | 101 | |
| GenBank mRNA | AF162685.1 | - | 10 - 11 | 1262 | 101 | |
| AK141422.1 | - | 9 - 10 | 1273 | 101 | ||
| AK161577.1 | - | 10 - 11 | 1268 | 101 | ||
| AK172253.1 | - | 10 - 11 | 1260 | 101 | ||
| BC053026.1 | - | 10 - 11 | 1275 | 5709543 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp2
Genomic Map