Assay Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178939.2 | NP_849270.1 | 2 - 3 | 219 | 66 | |
| GenBank mRNA | AK003410.1 | - | 2 - 3 | 221 | 66 | |
| AY286302.1 | - | 2 - 3 | 200 | 66 | ||
| BC031699.1 | - | 2 - 3 | 219 | 4020218 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdrg1
Genomic Map