Assay Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178939.2 | NP_849270.1 | 4 - 5 | 383 | 120 | |
| GenBank mRNA | AK003410.1 | - | 4 - 5 | 385 | 120 | |
| AK145939.1 | - | 4 - 5 | 428 | 120 | ||
| AY286302.1 | - | 4 - 5 | 364 | 120 | ||
| BC031699.1 | - | 4 - 5 | 383 | 4020218 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752415 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752417 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752418 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105010 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124613 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21752405 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124613 | - | - |
| Whole Mouse Genome G4122A | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | AK145939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | AY286302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | NM_178939.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P173961 | AK003410.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130080 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01677123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01388023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02154694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01677125 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02154696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752417 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752415 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21752418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21752421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02133690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01712917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02269263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02269260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02154696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02154694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01810546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01953803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02133689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02269258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01451272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01712916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02234893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01712919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01388023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21752403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01451273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488689 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428308_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428309_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419933_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488687 | - | - |
| Mouse Genome 430 2.0 Array | 1428309_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428308_at | - | - |
| Mouse Genome 430 2.0 Array | 1428308_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419934_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419934_at | - | - |
| Mouse Genome 430 2.0 Array | 1428309_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1419933_at | - | - |
| Mouse Genome 430 2.0 Array | 1419933_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68559_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68559_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041725.mm.1 | NM_178939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041725.mm.1 | NM_178939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041725.mm.1 | TC0200004922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041727.mm.1 | NM_178939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041727.mm.1 | NM_178939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041727.mm.1 | TC0200004922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041726.mm.1 | NM_178939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041726.mm.1 | NM_178939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041726.mm.1 | TC0200004922.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428308_at | - | - |
| Mouse Expression Set 430 | 1419933_PM_at | - | - |
| Mouse Expression Set 430 | 1419933_at | - | - |
| Mouse Expression Set 430 | 1428309_s_at | - | - |
| Mouse Expression Set 430 | 1428308_PM_at | - | - |
| Mouse Expression Set 430 | 1428309_PM_s_at | - | - |
| Murine Genome U74v2 Set | 92806_at | - | - |
| Murine Genome U74v2 Set | 92806_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419933_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428308_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428309_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428309_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419933_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428308_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428308_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419933_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428309_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419933_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428308_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428309_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 829551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5340592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249417 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249417 | - | - |
| mouse-6 v1.1 (Illumina) | scl068559.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068559.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249417 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249417 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068559.1_22-S | - | - |

Pdrg1
Genomic Map