Assay Details
| Gene Symbol: H13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159551.1 | NP_001153023.1 | 11 - 12 | 1155 | 72 | |
| GenBank mRNA | AK049101.1 | - | 11 - 12 | 1129 | 72 | |
| DQ168449.1 | - | 11 - 12 | 1171 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H13
Genomic Map