Assay Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011340.3 | NP_035470.3 | 4 - 5 | 570 | 94 | |
| GenBank mRNA | AF017057.1 | - | 4 - 5 | 468 | 94 | |
| BC019852.1 | - | 4 - 5 | 480 | 5123884 | 94 | |
| D50460.1 | - | 4 - 5 | 539 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinf1
Genomic Map