Assay Details
| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163336.1 | NP_001156808.1 | 5 - 6 | 649 | 67 | |
| NM_001163337.1 | NP_001156809.1 | 5 - 6 | 649 | 67 | ||
| NM_016745.3 | NP_058025.2 | 5 - 6 | 649 | 67 | ||
| GenBank mRNA | AK089487.1 | - | 5 - 6 | 649 | 67 | |
| AK155904.1 | - | 5 - 6 | 650 | 67 | ||
| BC017639.1 | - | 5 - 6 | 563 | 4924519 | 67 | |
| BC026147.1 | - | 5 - 6 | 595 | 4191458 | 67 | |
| U49393.1 | - | 5 - 6 | 510 | 67 | ||
| U49394.1 | - | 5 - 6 | 510 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp2a3
Genomic Map