Assay Details
| Gene Symbol: Ctns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031251.4 | NP_112541.1 | 9 - 10 | 969 | 79 | |
| GenBank mRNA | AJ250670.1 | - | 8 - 9 | 867 | 79 | |
| AK054467.1 | - | 10 - 11 | 1147 | 79 | ||
| AK078511.1 | - | 9 - 10 | 981 | 79 | ||
| AK079295.1 | - | 9 - 10 | 970 | 79 | ||
| AK085517.1 | - | 10 - 11 | 1150 | 79 | ||
| AK137143.1 | - | 9 - 10 | 1012 | 79 | ||
| AK170790.1 | - | 9 - 10 | 981 | 79 | ||
| AK170828.1 | - | 9 - 10 | 992 | 79 | ||
| BC005479.1 | - | 9 - 10 | 956 | 3484952 | 79 | |
| BC006033.1 | - | 9 - 10 | 956 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctns
Genomic Map