Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 12 - 13 | 1452 | 64 | |
| GenBank mRNA | AK028415.1 | - | 1 - 2 | 176 | 64 | |
| AK049281.1 | - | 12 - 13 | 1422 | 64 | ||
| AK085086.1 | - | 12 - 13 | 1406 | 64 | ||
| AK153905.1 | - | 12 - 13 | 1421 | 64 | ||
| AK208793.1 | - | 1 - 2 | 154 | 64 | ||
| AK213251.1 | - | 1 - 2 | 175 | 64 | ||
| AK214505.1 | - | 1 - 2 | 94 | 64 | ||
| BC006627.1 | - | 12 - 13 | 1457 | 3968733 | 64 | |
| BC019167.1 | - | 12 - 13 | 1405 | 4506351 | 64 | |
| U10120.1 | - | 12 - 13 | 1410 | 64 | ||
More Information
Set Membership: |
|

Nsf
Genomic Map