Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 13 - 14 | 1545 | 64 | |
| GenBank mRNA | AK028415.1 | - | 2 - 3 | 269 | 64 | |
| AK049281.1 | - | 13 - 14 | 1515 | 64 | ||
| AK085086.1 | - | 13 - 14 | 1499 | 64 | ||
| AK153905.1 | - | 13 - 14 | 1514 | 64 | ||
| AK208793.1 | - | 2 - 3 | 247 | 64 | ||
| AK213251.1 | - | 2 - 3 | 268 | 64 | ||
| AK214505.1 | - | 2 - 3 | 187 | 64 | ||
| BC006627.1 | - | 13 - 14 | 1550 | 3968733 | 64 | |
| BC019167.1 | - | 13 - 14 | 1498 | 4506351 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map