Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 1 - 2 | 82 | 77 | |
| GenBank mRNA | AK049281.1 | - | 1 - 2 | 52 | 77 | |
| AK051430.1 | - | 1 - 2 | 51 | 77 | ||
| AK141986.1 | - | 1 - 2 | 51 | 77 | ||
| AK153905.1 | - | 1 - 2 | 51 | 77 | ||
| BC006627.1 | - | 1 - 2 | 87 | 3968733 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049281.1, AK051430.1, AK153905.1, BC006627.1, NM_008740.4 transcripts.
Set Membership: |
|

Nsf
Genomic Map