Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 2 - 3 | 171 | 95 | |
| GenBank mRNA | AK042341.1 | - | 3 - 4 | 189 | 95 | |
| AK049281.1 | - | 2 - 3 | 141 | 95 | ||
| AK051430.1 | - | 2 - 3 | 140 | 95 | ||
| AK085086.1 | - | 2 - 3 | 125 | 95 | ||
| AK141986.1 | - | 2 - 3 | 140 | 95 | ||
| AK153905.1 | - | 2 - 3 | 140 | 95 | ||
| BC006627.1 | - | 2 - 3 | 176 | 3968733 | 95 | |
| BC019167.1 | - | 2 - 3 | 124 | 4506351 | 95 | |
| U10120.1 | - | 2 - 3 | 129 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map