Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 5 - 6 | 480 | 79 | |
| GenBank mRNA | AK042341.1 | - | 6 - 7 | 498 | 79 | |
| AK049281.1 | - | 5 - 6 | 450 | 79 | ||
| AK051430.1 | - | 5 - 6 | 449 | 79 | ||
| AK085086.1 | - | 5 - 6 | 434 | 79 | ||
| AK153905.1 | - | 5 - 6 | 449 | 79 | ||
| BC006627.1 | - | 5 - 6 | 485 | 3968733 | 79 | |
| BC019167.1 | - | 5 - 6 | 433 | 4506351 | 79 | |
| U10120.1 | - | 5 - 6 | 438 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map