Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 12 - 13 | 1603 | 79 | |
| GenBank mRNA | AK147084.1 | - | 12 - 13 | 1604 | 79 | |
| AK163950.1 | - | 2 - 3 | 291 | 79 | ||
| BC096493.1 | - | 12 - 13 | 1397 | 30742571 | 79 | |
| X52684.1 | - | 13 - 14 | 1544 | 79 | ||
| X52685.1 | - | 13 - 14 | 1544 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkca
Genomic Map